Gene Transcripts
- Gene Symbol
- DLST
- Entrez Gene ID
- 1743
- Gene Name
- dihydrolipoamide S-succinyltransferase
- Gene Aliases
- DLTS
- Chromosome Location
- Chr. 14: 74881891 - 74903747 on Build GRCh38
- UniGene ID
- Hs.525459
- Species
- Homo sapiens
- Gene Symbol
- DLSTP1
- Entrez Gene ID
- 1744
- Gene Name
- dihydrolipoamide S-succinyltransferase pseudogene 1
- Gene Aliases
- DLSTP
- Chromosome Location
- Chr. 1: 75742002 - 75745013 on Build GRCh38
- UniGene ID
- Hs.525459
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130749.1 |
|
1 |
363 |
|
GenBank mRNA |
AW834946.1 |
- |
1 |
434 |
|
S72422.1 |
- |
1 |
361 |
|
View Full Product Details