Gene Transcripts
- Gene Symbol
- FCER2
- Entrez Gene ID
- 2208
- Gene Name
- Fc fragment of IgE receptor II
- Gene Aliases
- BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
- Chromosome Location
- Chr. 19: 7688757 - 7702755 on Build GRCh38
- UniGene ID
- Hs.465778
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001207019.2 |
NP_001193948.2
|
6, 7 |
467 |
|
NM_001220500.1 |
NP_001207429.1
|
7, 8 |
603 |
|
NM_002002.4 |
NP_001993.2
|
7, 8 |
599 |
|
XM_005272462.4 |
XP_005272519.1
|
7, 8 |
1204 |
|
GenBank mRNA |
AK223207.1 |
- |
7, 8 |
568 |
|
AK310071.1 |
- |
6, 7 |
748 |
|
BC014108.2 |
- |
7, 8 |
564 |
4309266 |
BC062591.1 |
- |
7, 8 |
557 |
5216918 |
BC064417.1 |
- |
6, 7 |
881 |
|
M14766.1 |
- |
7, 8 |
571 |
|
M15059.1 |
- |
7, 8 |
599 |
|
X04772.1 |
- |
7, 8 |
599 |
|
View Full Product Details