Gene Transcripts

Gene Symbol
SCARF1
Entrez Gene ID
8578
Gene Name
scavenger receptor class F member 1
Gene Aliases
SREC, SREC-I, SREC1
Chromosome Location
Chr. 17: 1633858 - 1645789 on Build GRCh38
UniGene ID
Hs.647430
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003693.3 NP_003684.2 11 1883
NM_145350.2 NP_663325.1 11 1846
NR_028075.2 11 1813
NR_102409.1 11 1920
XM_017025256.1 XP_016880745.1 7 784
GenBank mRNA AB052947.1 - 11 1747
AB052948.1 - 11 1813
AB052949.1 - 10 2222
AB052950.1 - 9 1549
AK291810.1 - 11 1826
AK298737.1 - 8 965
AK310174.1 - 11 1843
BC039735.1 - 11 1851 6041360
D63483.1 - 11 1793
D86864.1 - 11 1817

View Full Product Details