Gene Transcripts

Gene Symbol
CHST2
Entrez Gene ID
9435
Gene Name
carbohydrate sulfotransferase 2
Gene Aliases
C6ST, GST-2, GST2, Gn6ST-1, HEL-S-75, glcNAc6ST-1
Chromosome Location
Chr. 3: 143119776 - 143124014 on Build GRCh38
UniGene ID
Hs.529249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004267.4 NP_004258.2 2 1925
GenBank mRNA AB014679.2 - 2 1234
AB014680.2 - 2 1234
AB021124.1 - 2 2370
AF083066.1 - 1 1043
AK023268.1 - 2 1875
BC017499.1 - 2 1606
BC036930.1 - 2 1836
BC068604.1 - 2 1875
BC105010.1 - 1 1005 8144013
BC105012.1 - 1 1005 8144015
EU668328.1 - 1 1005

View Full Product Details