Gene Transcripts

Gene Symbol
ICMT
Entrez Gene ID
23463
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
HSTE14, MST098, MSTP098, PCCMT, PCMT, PPMT
Chromosome Location
Chr. 1: 6221193 - 6235984 on Build GRCh38
UniGene ID
Hs.515688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012405.3 NP_036537.1 5 4093
XM_011541140.2 XP_011539442.1 4 3793
GenBank mRNA AF064084.1 - 6 2858
AL117548.1 - 2 1628

View Full Product Details