Gene Transcripts

Gene Symbol
A4GALT
Entrez Gene ID
53947
Gene Name
alpha 1,4-galactosyltransferase
Gene Aliases
A14GALT, A4GALT1, Gb3S, P(k), P1, P1PK, PK
Chromosome Location
Chr. 22: 42692115 - 42721301 on Build GRCh38
UniGene ID
Hs.105956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318038.1 NP_001304967.1 3 1370
NM_017436.5 NP_059132.1 3 1347
XM_005261644.2 XP_005261701.1 3 1652
XM_005261646.4 XP_005261703.1 2 1208
XM_005261647.2 XP_005261704.1 3 1182
XM_005261648.3 XP_005261705.1 3 1265
XM_006724265.3 XP_006724328.1 3 1593
XM_006724266.3 XP_006724329.1 3 1327
XM_011530233.2 XP_011528535.1 2 1534
XM_017028831.1 XP_016884320.1 2 1464
GenBank mRNA AB037883.1 - 2 1179
AJ245581.1 - 1 1046
AK074388.1 - 1 946
AK312927.1 - 3 1351
BC017068.2 - 3 1266 3913851
BC055286.1 - 2 1536 6161307
KU178649.1 - 1 1046
KU178650.1 - 2 665

View Full Product Details