Gene Transcripts

Gene Symbol
C1R
Entrez Gene ID
715
Gene Name
complement C1r subcomponent
Gene Aliases
Chromosome Location
Chr. 12: 7080209 - 7092447 on Build GRCh38
UniGene ID
Hs.524224
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001733.4 NP_001724.3 11 2304
GenBank mRNA AK024951.1 - 9 3310
AK222481.1 - 11 2298
BC035220.1 - 11 2304 4777940
M14058.1 - 11 2309
X04701.1 - 11 2296

View Full Product Details