Gene Transcripts
- Gene Symbol
- PGD
- Entrez Gene ID
- 5226
- Gene Name
- phosphogluconate dehydrogenase
- Gene Aliases
- 6PGD
- Chromosome Location
- Chr. 1: 10398992 - 10420511 on Build GRCh38
- UniGene ID
- Hs.464071
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304451.1 |
NP_001291380.1
|
6, 7 |
722 |
|
NM_001304452.1 |
NP_001291381.1
|
7, 8 |
867 |
|
NM_002631.3 |
NP_002622.2
|
7, 8 |
788 |
|
GenBank mRNA |
AK290404.1 |
- |
7, 8 |
713 |
|
AK296889.1 |
- |
8, 9 |
616 |
|
AK298830.1 |
- |
7, 8 |
867 |
|
AK300952.1 |
- |
6, 7 |
625 |
|
AK304423.1 |
- |
6, 7 |
645 |
|
BC000368.2 |
- |
7, 8 |
699 |
2819330 |
U30255.1 |
- |
6, 7 |
668 |
|
View Full Product Details