Gene Transcripts
- Gene Symbol
- TRPM2
- Entrez Gene ID
- 7226
- Gene Name
- transient receptor potential cation channel subfamily M member 2
- Gene Aliases
- EREG1, KNP3, LTRPC2, LTrpC-2, NUDT9H, NUDT9L1, TRPC7
- Chromosome Location
- Chr. 21: 44350112 - 44443081 on Build GRCh38
- UniGene ID
- Hs.369759
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320350.1 |
NP_001307279.1
|
1, 2 |
273 |
|
NM_001320351.1 |
NP_001307280.1
|
1, 2 |
273 |
|
NM_003307.3 |
NP_003298.1
|
1, 2 |
273 |
|
NR_038257.1 |
|
1, 2 |
273 |
|
XM_005261171.3 |
XP_005261228.1
|
2, 3 |
434 |
|
XM_011529736.2 |
XP_011528038.1
|
1, 2 |
677 |
|
XM_017028456.1 |
XP_016883945.1
|
2, 3 |
437 |
|
XM_017028457.1 |
XP_016883946.1
|
1, 2 |
678 |
|
GenBank mRNA |
AB001535.1 |
- |
1, 2 |
618 |
|
AB166745.1 |
- |
2, 3 |
386 |
|
AJ417076.1 |
- |
1, 2 |
186 |
|
AJ878416.1 |
- |
1, 2 |
236 |
|
AY603182.1 |
- |
1, 2 |
173 |
|
BC112342.1 |
- |
1, 2 |
186 |
40069893 |
View Full Product Details