Gene Transcripts

Gene Symbol
CAPN7
Entrez Gene ID
23473
Gene Name
calpain 7
Gene Aliases
CALPAIN7, PALBH
Chromosome Location
Chr. 3: 15206226 - 15252916 on Build GRCh38
UniGene ID
Hs.595234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014296.2 NP_055111.1 2 384
XM_011533555.2 XP_011531857.1 2 235
XM_011533556.2 XP_011531858.1 3 467
XM_011533557.2 XP_011531859.1 2 357
XM_017006043.1 XP_016861532.1 3 467
XM_017006044.1 XP_016861533.1 2 357
XM_017006045.1 XP_016861534.1 3 423
GenBank mRNA AB028639.1 - 2 365
AK314253.1 - 2 346
BC056202.1 - 2 362 5274889
BI464220.1 - 3 581

View Full Product Details