Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016113.4 | NP_057197.2 | 5 | 1067 | |
XM_005256676.1 | XP_005256733.1 | 5 | 1067 | ||
XM_005256677.1 | XP_005256734.1 | 5 | 1067 | ||
XM_005256678.4 | XP_005256735.1 | 5 | 1067 | ||
XM_006721541.3 | XP_006721604.1 | 5 | 1067 | ||
XM_006721543.3 | XP_006721606.1 | 5 | 1067 | ||
XM_011523922.1 | XP_011522224.1 | 5 | 1067 | ||
XM_011523923.1 | XP_011522225.1 | 5 | 1067 | ||
XM_017024730.1 | XP_016880219.1 | 5 | 1067 | ||
XM_017024731.1 | XP_016880220.1 | 5 | 1067 | ||
XM_017024732.1 | XP_016880221.1 | 5 | 1067 | ||
GenBank mRNA | AF103906.1 | - | 5 | 833 | |
AF129112.1 | - | 4 | 703 | ||
AJ487963.1 | - | 4 | 672 | ||
AK289705.1 | - | 5 | 1067 | ||
BC018926.2 | - | 5 | 1035 | 4298484 | |
BC051305.1 | - | 5 | 1023 | 4870010 |