Gene Transcripts

Gene Symbol
ZDHHC3
Entrez Gene ID
51304
Gene Name
zinc finger DHHC-type containing 3
Gene Aliases
DHHC-3, GODZ, ZNF373
Chromosome Location
Chr. 3: 44915261 - 44976190 on Build GRCh38
UniGene ID
Hs.61430
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135179.1 NP_001128651.1 7 2046
NM_016598.2 NP_057682.1 8 2130
XM_006713192.2 XP_006713255.1 9 2232
XM_006713193.2 XP_006713256.1 8 2149
XM_011533801.2 XP_011532103.1 7 2017
XM_017006566.1 XP_016862055.1 8 2084
XM_017006567.1 XP_016862056.1 8 2122
XM_017006569.1 XP_016862058.1 8 2121
GenBank mRNA AF052182.1 - 1 658
AF247703.1 - 8 2070
BC020350.1 - 1 824
BC020424.1 - 1 824
BC038436.1 - 1 824

View Full Product Details