Gene Transcripts

Gene Symbol
PROSER1
Entrez Gene ID
80209
Gene Name
proline and serine rich 1
Gene Aliases
C13orf23
Chromosome Location
Chr. 13: 39009865 - 39038115 on Build GRCh38
UniGene ID
Hs.318526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025138.4 NP_079414.3 11 1826
XM_005266545.3 XP_005266602.1 10 1799
XM_011535239.2 XP_011533541.1 10 1759
GenBank mRNA AB107354.1 - 11 1826
AK022723.1 - 5 502
AK074360.1 - 11 1798
AK299663.1 - 8 822
BC053566.1 - 5 505
BC064348.1 - 11 1081 4939198
BX647471.1 - 11 1248

View Full Product Details