Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136050.2 | NP_001129522.1 | 6, 7 | 936 | |
NM_138452.2 | NP_612461.1 | 6, 7 | 928 | ||
GenBank mRNA | AF418205.1 | - | 6, 7 | 872 | |
AK058159.1 | - | 6, 7 | 928 | ||
AL833917.1 | - | 6, 7 | 831 | ||
BC014057.2 | - | 6, 7 | 842 | 3622913 | |
BC015943.1 | - | 6, 7 | 739 | 3922816 | |
BX247980.1 | - | 6, 7 | 852 |