Gene Transcripts
- Gene Symbol
- PPP6C
- Entrez Gene ID
- 5537
- Gene Name
- protein phosphatase 6 catalytic subunit
- Gene Aliases
- PP6, PP6C
- Chromosome Location
- Chr. 9: 125146573 - 125189939 on Build GRCh38
- UniGene ID
- Hs.744091
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001123369.1 |
NP_001116841.1
|
1, 2 |
295 |
|
NM_002721.4 |
NP_002712.1
|
1, 2 |
295 |
|
GenBank mRNA |
AF035158.2 |
- |
1, 2 |
74 |
|
AK295752.1 |
- |
1, 2 |
115 |
|
AK299087.1 |
- |
1, 2 |
159 |
|
AK312332.1 |
- |
1, 2 |
127 |
|
AK316527.1 |
- |
1, 2 |
149 |
|
BC006990.1 |
- |
1, 2 |
142 |
3684339 |
BT019708.1 |
- |
1, 2 |
74 |
|
DB037210.1 |
- |
1, 2 |
295 |
|
DC369815.1 |
- |
1, 2 |
159 |
|
KU178271.1 |
- |
1, 2 |
74 |
|
KU178272.1 |
- |
1, 2 |
74 |
|
X92972.2 |
- |
1, 2 |
95 |
|
View Full Product Details