Gene Transcripts

Gene Symbol
MTMR1
Entrez Gene ID
8776
Gene Name
myotubularin related protein 1
Gene Aliases
Chromosome Location
Chr. X: 150692997 - 150765103 on Build GRCh38
UniGene ID
Hs.347187
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306144.1 NP_001293073.1 15 1872
NM_003828.3 NP_003819.1 14 1848
XM_005274765.3 XP_005274822.1 17 1912
XM_005274766.3 XP_005274823.1 16 1860
XM_005274767.3 XP_005274824.1 15 1879
XM_006724855.3 XP_006724918.1 14 1848
XM_011531209.2 XP_011529511.1 14 1608
XM_017029921.1 XP_016885410.1 15 1643
XM_017029922.1 XP_016885411.1 15 1655
XM_017029923.1 XP_016885412.1 12 1435
GenBank mRNA AF057354.1 - 15 1883
AJ224979.1 - 14 1674
AK001816.1 - 12 1402
AK226109.1 - 15 1826
AK295738.1 - 15 1667
U58032.1 - 11 1453

View Full Product Details