Gene Transcripts
- Gene Symbol
- RAB4A
- Entrez Gene ID
- 5867
- Gene Name
- RAB4A, member RAS oncogene family
- Gene Aliases
- HRES-1, HRES-1/RAB4, HRES1, RAB4
- Chromosome Location
- Chr. 1: 229271062 - 229305894 on Build GRCh38
- UniGene ID
- Hs.296169
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004578.3 |
NP_004569.2
|
3, 4 |
506 |
|
NR_073545.1 |
|
3, 4 |
506 |
|
GenBank mRNA |
AF498934.1 |
- |
3, 4 |
228 |
|
AK223009.1 |
- |
3, 4 |
271 |
|
AK313807.1 |
- |
3, 4 |
479 |
|
AY585832.1 |
- |
3, 4 |
435 |
|
BC002438.1 |
- |
3, 4 |
299 |
3346455 |
BC004309.1 |
- |
3, 4 |
436 |
3627110 |
BI828974.1 |
- |
3, 4 |
390 |
|
KC407675.1 |
- |
3, 4 |
228 |
|
M28211.1 |
- |
3, 4 |
283 |
|
X13902.1 |
- |
3, 4 |
339 |
|
View Full Product Details