Gene Transcripts

Gene Symbol
LUM
Entrez Gene ID
4060
Gene Name
lumican
Gene Aliases
LDC, SLRR2D
Chromosome Location
Chr. 12: 91103455 - 91111765 on Build GRCh38
UniGene ID
Hs.406475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002345.3 NP_002336.1 2 722
GenBank mRNA AK223177.1 - 2 407
AK312682.1 - 2 425
BC007038.1 - 2 474 3950745
BC035997.1 - 2 396 4688606
BT006707.1 - 1 333
U18728.1 - 2 413
U21128.1 - 2 417

View Full Product Details