Gene Transcripts

Gene Symbol
PWP1
Entrez Gene ID
11137
Gene Name
PWP1 homolog, endonuclein
Gene Aliases
IEF-SSP-9502
Chromosome Location
Chr. 12: 107685732 - 107713162 on Build GRCh38
UniGene ID
Hs.506652
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317962.1 NP_001304891.1 15 1524
NM_001317963.1 NP_001304892.1 15 1696
NM_007062.2 NP_008993.1 15 1668
GenBank mRNA AK223162.1 - 15 1592
AK290681.1 - 15 1509
AK296252.1 - 15 1525
AK297961.1 - 14 1353
BC001652.2 - 15 1587 2924215
L07758.1 - 15 1587

View Full Product Details