Gene Transcripts
- Gene Symbol
- RRN3P1
- Entrez Gene ID
- 730092
- Gene Name
- RRN3 homolog, RNA polymerase I transcription factor pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 16: 21796630 - 21819174 on Build GRCh38
- UniGene ID
- Hs.31290
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003370.2 |
|
12 |
1493 |
|
GenBank mRNA |
BC006441.1 |
- |
6 |
2492 |
|
View Full Product Details