Gene Transcripts

Gene Symbol
SLC7A2
Entrez Gene ID
6542
Gene Name
solute carrier family 7 member 2
Gene Aliases
ATRC2, CAT2, HCAT2
Chromosome Location
Chr. 8: 17494349 - 17570568 on Build GRCh38
UniGene ID
Hs.448520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008539.3 NP_001008539.3 5 886
NM_001164771.1 NP_001158243.1 5 882
NM_003046.5 NP_003037.4 5 882
XM_005273610.4 XP_005273667.1 6 947
XM_005273611.4 XP_005273668.1 6 850
XM_005273612.4 XP_005273669.1 6 944
XM_017013746.1 XP_016869235.1 5 885
XM_017013747.1 XP_016869236.1 6 947
GenBank mRNA AK314287.1 - 5 884
BC069648.1 - 5 789 7262473
BC104905.1 - 5 775 8143908
BC113661.1 - 5 775 8322713
BC143583.1 - 5 775 9052094
D29990.1 - 4 714
U76368.1 - 5 908

View Full Product Details