Gene Transcripts

Gene Symbol
STAT4
Entrez Gene ID
6775
Gene Name
signal transducer and activator of transcription 4
Gene Aliases
SLEB11
Chromosome Location
Chr. 2: 191029576 - 191172684 on Build GRCh38
UniGene ID
Hs.735572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243835.1 NP_001230764.1 3 500
NM_003151.3 NP_003142.1 3 551
XM_006712719.3 XP_006712782.1 3 546
XM_011511705.2 XP_011510007.1 6 697
XM_017004784.1 XP_016860273.1 3 305
GenBank mRNA AK299976.1 - 3 272
AK300868.1 - 3 304
BC031212.1 - 3 472 4830583
BF966132.1 - 3 498
EU304788.1 - 4 515
EU304789.1 - 3 859
EU304792.1 - 4 540
EU304793.1 - 3 1156
L78440.1 - 3 317

View Full Product Details