Gene Transcripts

Gene Symbol
TULP1
Entrez Gene ID
7287
Gene Name
tubby like protein 1
Gene Aliases
LCA15, RP14, TUBL1
Chromosome Location
Chr. 6: 35497874 - 35512902 on Build GRCh38
UniGene ID
Hs.485208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289395.1 NP_001276324.1 5 425
NM_003322.4 NP_003313.3 6 584
GenBank mRNA AB593123.1 - 6 584
AB593124.1 - 6 570
BC032714.1 - 5 410 5495136
BC065261.1 - 5 393 6144003
CN355966.1 - 2 393
U82468.1 - 6 576

View Full Product Details