Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014564.4 | NP_055379.1 | 6 | 1249 | |
NM_178138.5 | NP_835258.1 | 6 | 1217 | ||
XM_005263410.1 | XP_005263467.1 | 6 | 1531 | ||
XM_017015168.1 | XP_016870657.1 | 5 | 1028 | ||
GenBank mRNA | AB593062.1 | - | 6 | 1217 | |
AB593063.1 | - | 6 | 1216 | ||
AB593064.1 | - | 6 | 1531 | ||
AB593065.1 | - | 5 | 1925 | ||
AB593066.1 | - | 6 | 1249 | ||
AB593067.1 | - | 6 | 1213 | ||
AF096169.1 | - | 5 | 1021 | ||
AF156888.1 | - | 6 | 1204 | ||
AF156889.1 | - | 6 | 1234 |