Gene Transcripts

Gene Symbol
RNF125
Entrez Gene ID
54941
Gene Name
ring finger protein 125
Gene Aliases
TNORS, TRAC-1, TRAC1
Chromosome Location
Chr. 18: 32018203 - 32090751 on Build GRCh38
UniGene ID
Hs.633703
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017831.3 NP_060301.2 1, 2 552
XM_011526045.2 XP_011524347.1 1, 2 821
XM_011526046.2 XP_011524348.1 1, 2 202
XM_011526047.2 XP_011524349.1 1, 2 831
GenBank mRNA AK000463.1 - 1, 2 552
AK225518.1 - 1, 2 552
BC012021.1 - 1, 2 183 4521247
BI666910.1 - 1, 2 667
BM469263.1 - 1, 2 183

View Full Product Details