Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242894.1 | NP_001229823.1 | 7 | 1160 | |
NM_001242895.1 | NP_001229824.1 | 5 | 579 | ||
NM_024493.3 | NP_077819.2 | 6 | 1043 | ||
XM_005249423.3 | XP_005249480.1 | 5 | 1209 | ||
XM_006715215.2 | XP_006715278.1 | 6 | 941 | ||
XM_006715216.2 | XP_006715279.1 | 6 | 956 | ||
XM_006715218.3 | XP_006715281.1 | 5 | 507 | ||
XM_017011320.1 | XP_016866809.1 | 7 | 1037 | ||
GenBank mRNA | AK091225.1 | - | 4 | 2253 | |
AK122790.1 | - | 5 | 543 | ||
AK313532.1 | - | 7 | 1124 | ||
BC006118.1 | - | 6 | 975 | 3505521 | |
BT007427.1 | - | 5 | 827 | ||
U71601.1 | - | 3 | 214 |