Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024915.3 | NP_079191.2 | 4 | 637 | |
XM_011517305.2 | XP_011515607.1 | 4 | 668 | ||
XM_011517306.2 | XP_011515608.1 | 4 | 405 | ||
XM_011517307.2 | XP_011515609.1 | 4 | 635 | ||
GenBank mRNA | AK023844.1 | - | 4 | 319 | |
AK292853.1 | - | 4 | 319 | ||
AK296817.1 | - | 4 | 636 | ||
BC069618.1 | - | 4 | 319 | 7262389 | |
BC069633.1 | - | 4 | 379 | 7262401 | |
BC069638.1 | - | 4 | 319 | 7262413 | |
BC129822.1 | - | 4 | 319 | 40113571 | |
BC129823.1 | - | 4 | 319 | 40113576 |