Gene Transcripts
- Gene Symbol
- HGD
- Entrez Gene ID
- 3081
- Gene Name
- homogentisate 1,2-dioxygenase
- Gene Aliases
- AKU, HGO
- Chromosome Location
- Chr. 3: 120628168 - 120682571 on Build GRCh38
- UniGene ID
- Hs.368254
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000187.3 |
NP_000178.2
|
4, 5 |
750 |
|
XM_005247412.1 |
XP_005247469.1
|
4, 5 |
750 |
|
XM_005247413.1 |
XP_005247470.1
|
4, 5 |
750 |
|
XM_005247414.4 |
XP_005247471.1
|
4, 5 |
750 |
|
XM_011512746.1 |
XP_011511048.1
|
4, 5 |
750 |
|
XM_017006277.1 |
XP_016861766.1
|
4, 5 |
363 |
|
GenBank mRNA |
AF045167.1 |
- |
4, 5 |
457 |
|
AK290782.1 |
- |
4, 5 |
456 |
|
AK313563.1 |
- |
4, 5 |
734 |
|
BC020792.1 |
- |
4, 5 |
456 |
4716316 |
BC071757.1 |
- |
4, 5 |
388 |
6714217 |
U63008.1 |
- |
4, 5 |
419 |
|
Z75048.1 |
- |
4, 5 |
419 |
|
View Full Product Details