Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289050.1 | NP_001275979.1 | 9 | 1895 | |
NM_016609.4 | NP_057693.3 | 10 | 2287 | ||
NM_020372.3 | NP_065105.2 | 9 | 2074 | ||
NR_110290.1 | 8 | 1952 | |||
XM_005267747.4 | XP_005267804.1 | 10 | 4417 | ||
XM_005267748.4 | XP_005267805.1 | 8 | 4042 | ||
XM_017021361.1 | XP_016876850.1 | 10 | 4414 | ||
XM_017021362.1 | XP_016876851.1 | 10 | 4358 | ||
GenBank mRNA | AJ243653.1 | - | 7 | 1335 | |
BC020565.1 | - | 9 | 1893 | 4156259 | |
BC090870.1 | - | 9 | 1571 | ||
BC111015.1 | - | 8 | 1434 | ||
BF345519.1 | - | 1 | 84 | ||
BX161416.1 | - | 10 | 2246 | ||
BX647316.1 | - | 6 | 1642 |