Gene Transcripts

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
Chromosome Location
Chr. 17: 11598431 - 11970168 on Build GRCh38
UniGene ID
Hs.567259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001372.3 NP_001363.2 59, 60 11474
NM_004662.2 NP_004653.2 5, 6 871
XM_011523703.1 XP_011522005.1 59, 60 11438
XM_017024292.1 XP_016879781.1 58, 59 11321
XM_017024293.1 XP_016879782.1 48, 49 9382
GenBank mRNA AB002355.2 - 40, 41 7152
AB290158.1 - 47, 48 9291
AF257737.1 - 59, 60 11464
AJ404468.1 - 59, 60 11474
AK302009.1 - 4, 5 687
BC128261.1 - 3, 4 411
BC128421.1 - 3, 4 411
BC153883.1 - 40, 41 7152
X99947.1 - 5, 6 871

View Full Product Details