Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204514.1 | NP_001191443.1 | 2 | 355 | |
NM_001204517.1 | NP_001191446.1 | 2 | 1464 | ||
NM_014970.3 | NP_055785.2 | 2 | 355 | ||
XM_005244970.1 | XP_005245027.1 | 2 | 358 | ||
XM_011509307.2 | XP_011507609.2 | 2 | 358 | ||
GenBank mRNA | AK122897.1 | - | 2 | 1466 | |
AK303052.1 | - | 2 | 312 | ||
AK315586.1 | - | 2 | 269 | ||
BC028679.2 | - | 2 | 312 | 4829354 | |
BG707718.1 | - | 2 | 364 | ||
DA144269.1 | - | 2 | 358 | ||
U59919.1 | - | 2 | 272 |