Gene Transcripts
- Gene Symbol
- CCNE1
- Entrez Gene ID
- 898
- Gene Name
- cyclin E1
- Gene Aliases
- CCNE, pCCNE1
- Chromosome Location
- Chr. 19: 29811994 - 29824317 on Build GRCh38
- UniGene ID
- Hs.244723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001238.3 |
NP_001229.1
|
6, 7 |
653 |
|
NM_001322259.1 |
NP_001309188.1
|
6, 7 |
653 |
|
NM_001322262.1 |
NP_001309191.1
|
5, 6 |
606 |
|
XM_011527440.2 |
XP_011525742.1
|
6, 7 |
539 |
|
GenBank mRNA |
AK291549.1 |
- |
6, 7 |
653 |
|
BC035498.2 |
- |
6, 7 |
643 |
3841192 |
KF672847.1 |
- |
4, 5 |
482 |
|
KR134287.1 |
- |
6, 7 |
653 |
|
M73812.1 |
- |
4, 5 |
482 |
|
M74093.1 |
- |
4, 5 |
450 |
|
View Full Product Details