Gene Transcripts

Gene Symbol
NIT1
Entrez Gene ID
4817
Gene Name
nitrilase 1
Gene Aliases
Chromosome Location
Chr. 1: 161118072 - 161125446 on Build GRCh38
UniGene ID
Hs.146406
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185092.1 NP_001172021.1 5, 6 696
NM_001185093.1 NP_001172022.1 4, 5 928
NM_001185094.1 NP_001172023.1 5, 6 697
NM_005600.2 NP_005591.1 5, 6 696
XM_005245214.2 XP_005245271.1 5, 6 1047
XM_005245215.4 XP_005245272.1 5, 6 708
XM_005245216.3 XP_005245273.1 5, 6 719
XM_006711341.3 XP_006711404.1 5, 6 707
XM_017001371.1 XP_016856860.1 5, 6 723
GenBank mRNA AF069987.1 - 5, 6 667
AK296606.1 - 4, 5 892
AK313323.1 - 5, 6 663
BC046149.1 - 5, 6 650 5764281
BX416745.2 - 5, 6 663
CR541814.1 - 4, 5 591
CR541846.1 - 4, 5 591

View Full Product Details