Gene Transcripts

Gene Symbol
ART4
Entrez Gene ID
420
Gene Name
ADP-ribosyltransferase 4 (Dombrock blood group)
Gene Aliases
ARTC4, CD297, DO, DOK1
Chromosome Location
Chr. 12: 14829311 - 14843479 on Build GRCh38
UniGene ID
Hs.591158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021071.2 NP_066549.2 2 672
GenBank mRNA AF290204.1 - 2 405
AK291662.1 - 2 688
AY829434.1 - 2 641
AY829437.1 - 2 255
BC074727.2 - 2 342 30915180
BX096478.1 - 1 60
CR746266.1 - 2 672

View Full Product Details