Gene Transcripts

Gene Symbol
ADAT1
Entrez Gene ID
23536
Gene Name
adenosine deaminase, tRNA specific 1
Gene Aliases
HADAT1
Chromosome Location
Chr. 16: 75596879 - 75623323 on Build GRCh38
UniGene ID
Hs.729312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324444.1 NP_001311373.1 3 283
NM_001324445.1 NP_001311374.1 2 1056
NM_001324446.1 NP_001311375.1 2 962
NM_001324448.1 NP_001311377.1 3 283
NM_001324449.1 NP_001311378.1 2 1056
NM_001324450.1 NP_001311379.1 3 283
NM_001324451.1 NP_001311380.1 2 1056
NM_001324452.1 NP_001311381.1 2 1056
NM_001324453.1 NP_001311382.1 2 1056
NM_012091.4 NP_036223.2 3 283
GenBank mRNA AF125188.1 - 2 163
AK001687.1 - 3 216
AK296834.1 - 4 154
AK314652.1 - 3 225
BC002758.2 - 3 229 3632079
DC395938.1 - 3 283

View Full Product Details