Gene Transcripts

Gene Symbol
SLC39A1
Entrez Gene ID
27173
Gene Name
solute carrier family 39 member 1
Gene Aliases
ZIP1, ZIRTL
Chromosome Location
Chr. 1: 153959099 - 153968184 on Build GRCh38
UniGene ID
Hs.743291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271957.1 NP_001258886.1 4 1453
NM_001271958.1 NP_001258887.1 4 1290
NM_001271959.1 NP_001258888.1 4 1459
NM_001271960.1 NP_001258889.1 5 1351
NM_001271961.1 NP_001258890.1 3 1159
NM_014437.4 NP_055252.2 5 1597
GenBank mRNA AF132942.1 - 3 1147
AF151829.1 - 4 1168
AJ243649.1 - 4 1184
AJ271671.1 - 4 1198
AK074943.1 - 4 1407
AK075257.1 - 4 1221
AK293389.1 - 3 1067
AK297983.1 - 5 1165
BC002563.2 - 4 1172 3138575
BC003152.2 - 3 1144 2958651
BC007886.2 - 4 1169 4122086
BC014303.2 - 5 1577 3942586

View Full Product Details