Gene Transcripts
- Gene Symbol
- PPP1R10
- Entrez Gene ID
- 5514
- Gene Name
- protein phosphatase 1 regulatory subunit 10
- Gene Aliases
- CAT53, FB19, PNUTS, PP1R10, R111, p99
- Chromosome Location
- Chr. 6: 30600400 - 30618607 on Build GRCh38
- UniGene ID
- Hs.106019
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002714.3 |
NP_002705.2
|
6, 7 |
999 |
|
NR_072994.1 |
|
6, 7 |
999 |
|
XM_006715130.2 |
XP_006715193.1
|
6, 7 |
965 |
|
XM_011514722.1 |
XP_011513024.1
|
7, 8 |
989 |
|
XM_017010995.1 |
XP_016866484.1
|
5, 6 |
1134 |
|
GenBank mRNA |
AJ544537.1 |
- |
6, 7 |
932 |
|
AK307406.1 |
- |
6, 7 |
935 |
|
BC005267.2 |
- |
5, 6 |
916 |
|
BC136294.1 |
- |
5, 6 |
583 |
9020281 |
BC150310.1 |
- |
6, 7 |
1000 |
|
Y13247.1 |
- |
5, 6 |
922 |
|
View Full Product Details