Gene Transcripts

Gene Symbol
ATP6V1E1
Entrez Gene ID
529
Gene Name
ATPase H+ transporting V1 subunit E1
Gene Aliases
ATP6E, ATP6E2, ATP6V1E, P31, Vma4
Chromosome Location
Chr. 22: 17592136 - 17628822 on Build GRCh38
UniGene ID
Hs.517338
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039366.1 NP_001034455.1 3 383
NM_001039367.1 NP_001034456.1 4 449
NM_001696.3 NP_001687.1 4 449
GenBank mRNA AK294623.1 - 3 310
AK312394.1 - 4 370
AK315941.1 - 4 375
BC004443.1 - 4 351 2820930
BE735148.1 - 4 395
BI546032.1 - 4 401
BI597419.1 - 4 379
BQ888820.1 - 3 328
BT007128.1 - 4 262
CK005516.1 - 4 452
CR456385.1 - 4 315
X71491.1 - 4 318
X76228.1 - 4 337

View Full Product Details