Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142930.1 | NP_001136402.1 | 5 | 613 | |
NM_001142931.1 | NP_001136403.1 | 4 | 451 | ||
NM_006595.3 | NP_006586.1 | 5 | 613 | ||
NR_024625.1 | 6 | 675 | |||
XM_006718359.3 | XP_006718422.1 | 5 | 603 | ||
XM_017018464.1 | XP_016873953.1 | 6 | 660 | ||
XM_017018465.1 | XP_016873954.1 | 6 | 660 | ||
GenBank mRNA | AF229253.1 | - | 4 | 227 | |
AF229254.1 | - | 5 | 553 | ||
AK294724.1 | - | 4 | 408 | ||
AK299288.1 | - | 5 | 579 | ||
AK304157.1 | - | 6 | 641 | ||
AY311389.1 | - | 5 | 552 | ||
BC017709.1 | - | 5 | 533 | 4419401 | |
BT007093.1 | - | 5 | 440 | ||
U83857.1 | - | 5 | 535 | ||
Y15906.1 | - | 5 | 572 |