Gene Transcripts

Gene Symbol
ARHGEF26
Entrez Gene ID
26084
Gene Name
Rho guanine nucleotide exchange factor 26
Gene Aliases
CSGEF, HMFN1864, SGEF
Chromosome Location
Chr. 3: 154121003 - 154257827 on Build GRCh38
UniGene ID
Hs.240845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251962.1 NP_001238891.1 15 3617
NM_001251963.1 NP_001238892.1 15 3479
NM_015595.3 NP_056410.3 15 3544
XM_011512672.1 XP_011510974.1 14 3454
GenBank mRNA AF415175.1 - 15 3499
AF415176.1 - 5 1469
AK022655.1 - 9 1855
AK022884.1 - 12 2131
AK123040.1 - 15 3423
AK127282.1 - 15 3617
AL117429.1 - 5 1355
BC016628.1 - 9 1761
BC078655.1 - 15 3485 4374088

View Full Product Details