Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020376.3 | NP_065109.1 | 3 | 578 | |
XM_006718265.3 | XP_006718328.1 | 3 | 584 | ||
XM_006718266.3 | XP_006718329.1 | 2 | 524 | ||
XM_017018028.1 | XP_016873517.1 | 2 | 524 | ||
GenBank mRNA | AF055000.1 | - | 1 | 96 | |
AJ278475.1 | - | 3 | 447 | ||
AJ278476.1 | - | 2 | 467 | ||
AK308435.1 | - | 2 | 544 | ||
AY203925.1 | - | 1 | 982 | ||
AY894804.1 | - | 2 | 375 | ||
BC017280.2 | - | 3 | 577 | 4875483 | |
HQ651812.1 | - | 3 | 578 | ||
JF279441.1 | - | 3 | 578 | ||
JF279442.1 | - | 3 | 578 | ||
KF601369.1 | - | 3 | 578 | ||
KF833264.1 | - | 2 | 375 | ||
KF833265.1 | - | 2 | 375 |