Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000034.3 | NP_000025.1 | 12 | 1752 | |
NM_001127617.2 | NP_001121089.1 | 7 | 974 | ||
NM_001243177.1 | NP_001230106.1 | 8 | 1095 | ||
NM_184041.2 | NP_908930.1 | 7 | 941 | ||
NM_184043.2 | NP_908932.1 | 7 | 913 | ||
XM_011545768.2 | XP_011544070.1 | 9 | 1615 | ||
GenBank mRNA | AK026577.1 | - | 7 | 790 | |
AK098778.1 | - | 10 | 1793 | ||
AK301993.1 | - | 8 | 907 | ||
BC000367.2 | - | 4 | 353 | ||
BC004333.1 | - | 12 | 1672 | 3630974 | |
BC010660.1 | - | 7 | 859 | 3869603 | |
BC012880.1 | - | 7 | 787 | 3459009 | |
BC013614.1 | - | 7 | 870 | 3880857 | |
BC015888.2 | - | 7 | 783 | 3911623 | |
BC016170.1 | - | 4 | 353 | ||
BC016800.1 | - | 8 | 811 | 4072487 | |
BU855133.1 | - | 2 | 60 | ||
BX647566.1 | - | 7 | 763 | ||
CR536528.1 | - | 6 | 665 | ||
CR541880.1 | - | 6 | 665 | ||
EF036496.1 | - | 4 | 313 | ||
EF036497.1 | - | 3 | 244 | ||
FJ474908.1 | - | 8 | 832 | ||
M11560.1 | - | 8 | 832 | ||
M21190.1 | - | 3 | 251 | ||
X05236.1 | - | 7 | 811 |