Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000846.4 | NP_000837.3 | 5 | 498 | |
XM_011514532.2 | XP_011512834.1 | 5 | 490 | ||
GenBank mRNA | AK292602.1 | - | 5 | 410 | |
AK313601.1 | - | 5 | 410 | ||
BC002895.2 | - | 5 | 461 | 3943417 | |
BE795593.1 | - | 5 | 441 | ||
BE796134.1 | - | 5 | 442 | ||
M16594.1 | - | 5 | 414 |