Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286233.1 | NP_001273162.1 | 16 | 3113 | |
NM_001286234.1 | NP_001273163.1 | 11 | 2359 | ||
NM_001286235.1 | NP_001273164.1 | 13 | 2657 | ||
NM_001286236.1 | NP_001273165.1 | 10 | 2198 | ||
NM_001286237.1 | NP_001273166.1 | 10 | 2252 | ||
NM_153449.3 | NP_703150.1 | 12 | 2395 | ||
XM_005253315.3 | XP_005253372.1 | 11 | 2193 | ||
XM_005253317.4 | XP_005253374.1 | 11 | 2712 | ||
XM_011520562.1 | XP_011518864.1 | 14 | 2547 | ||
XM_011520563.2 | XP_011518865.1 | 11 | 2207 | ||
XM_011520564.2 | XP_011518866.1 | 10 | 2016 | ||
XM_011520565.2 | XP_011518867.1 | 9 | 2045 | ||
XM_017018841.1 | XP_016874330.1 | 12 | 2292 | ||
XM_017018842.1 | XP_016874331.1 | 15 | 2863 | ||
XM_017018843.1 | XP_016874332.1 | 15 | 3187 | ||
XM_017018844.1 | XP_016874333.1 | 12 | 2242 | ||
XM_017018845.1 | XP_016874334.1 | 12 | 2784 | ||
XM_017018846.1 | XP_016874335.1 | 12 | 2368 | ||
XM_017018847.1 | XP_016874336.1 | 11 | 2325 | ||
GenBank mRNA | AK122783.1 | - | 13 | 2444 | |
AK126026.1 | - | 16 | 2894 | ||
AL110298.1 | - | 5 | 1438 | ||
AL832448.1 | - | 9 | 2018 | ||
AY357941.1 | - | 11 | 2338 | ||
BC060766.1 | - | 11 | 2206 | 5297510 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006931.2 | NP_008862.1 | 10 | 2412 | |
GenBank mRNA | AB209607.1 | - | 9 | 2890 | |
AK074861.1 | - | 2 | 1050 | ||
M20681.1 | - | 10 | 2392 |