Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018837.1 | NP_001018238.1 | 2 | 274 | |
NM_006118.3 | NP_006109.2 | 2 | 418 | ||
GenBank mRNA | AK290626.1 | - | 2 | 340 | |
AK294298.1 | - | 2 | 344 | ||
AK300676.1 | - | 2 | 372 | ||
AK310000.1 | - | 2 | 338 | ||
AK313291.1 | - | 2 | 349 | ||
BC005240.1 | - | 2 | 470 | 3931805 | |
BC014314.1 | - | 2 | 339 | 3931852 | |
BC015209.1 | - | 2 | 311 | 3883802 | |
BC016730.1 | - | 2 | 356 | 4082810 | |
BF668367.1 | - | 249 | |||
EU190982.1 | - | 2 | 352 | ||
EU190983.1 | - | 2 | 335 | ||
KU178523.1 | - | 2 | 257 | ||
KU178524.1 | - | 2 | 113 | ||
U68566.1 | - | 2 | 418 |