Gene Transcripts
- Gene Symbol
- UBAP2L
- Entrez Gene ID
- 9898
- Gene Name
- ubiquitin associated protein 2 like
- Gene Aliases
- NICE-4, NICE4
- Chromosome Location
- Chr. 1: 154220172 - 154271510 on Build GRCh38
- UniGene ID
- Hs.490551
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127320.2 |
NP_001120792.1
|
7, 8 |
704 |
|
NM_001287815.1 |
NP_001274744.1
|
7, 8 |
737 |
|
NM_001287816.1 |
NP_001274745.1
|
7, 8 |
791 |
|
NM_014847.3 |
NP_055662.3
|
7, 8 |
758 |
|
XM_005245658.3 |
XP_005245715.1
|
7, 8 |
791 |
|
XM_005245667.3 |
XP_005245724.1
|
7, 8 |
791 |
|
XM_005245668.3 |
XP_005245725.1
|
7, 8 |
791 |
|
XM_005245669.3 |
XP_005245726.1
|
7, 8 |
791 |
|
XM_005245670.3 |
XP_005245727.1
|
7, 8 |
791 |
|
XM_005245672.3 |
XP_005245729.1
|
7, 8 |
683 |
|
XM_005245673.3 |
XP_005245730.1
|
7, 8 |
791 |
|
XM_005245674.1 |
XP_005245731.1
|
7, 8 |
683 |
|
XM_011510215.2 |
XP_011508517.1
|
7, 8 |
791 |
|
XM_011510216.1 |
XP_011508518.1
|
7, 8 |
683 |
|
XM_017002972.1 |
XP_016858461.1
|
7, 8 |
816 |
|
XM_017002973.1 |
XP_016858462.1
|
7, 8 |
683 |
|
XM_017002974.1 |
XP_016858463.1
|
7, 8 |
714 |
|
XM_017002975.1 |
XP_016858464.1
|
7, 8 |
791 |
|
XM_017002976.1 |
XP_016858465.1
|
7, 8 |
693 |
|
XM_017002977.1 |
XP_016858466.1
|
7, 8 |
683 |
|
XM_017002978.1 |
XP_016858467.1
|
7, 8 |
714 |
|
XM_017002979.1 |
XP_016858468.1
|
7, 8 |
675 |
|
XM_017002980.1 |
XP_016858469.1
|
7, 8 |
714 |
|
XM_017002981.1 |
XP_016858470.1
|
7, 8 |
675 |
|
XM_017002982.1 |
XP_016858471.1
|
7, 8 |
683 |
|
XM_017002983.1 |
XP_016858472.1
|
7, 8 |
714 |
|
XM_017002984.1 |
XP_016858473.1
|
7, 8 |
714 |
|
XM_017002985.1 |
XP_016858474.1
|
7, 8 |
816 |
|
XM_017002986.1 |
XP_016858475.1
|
7, 8 |
683 |
|
XM_017002987.1 |
XP_016858476.1
|
7, 8 |
714 |
|
XM_017002988.1 |
XP_016858477.1
|
7, 8 |
714 |
|
XM_017002989.1 |
XP_016858478.1
|
7, 8 |
714 |
|
XM_017002990.1 |
XP_016858479.1
|
7, 8 |
675 |
|
XM_017002991.1 |
XP_016858480.1
|
6, 7 |
616 |
|
GenBank mRNA |
AK124294.1 |
- |
8, 9 |
613 |
|
AK302662.1 |
- |
7, 8 |
725 |
|
AK302953.1 |
- |
4, 5 |
452 |
|
AK303533.1 |
- |
7, 8 |
664 |
|
BC003170.1 |
- |
6, 7 |
640 |
2906083 |
D63478.1 |
- |
7, 8 |
697 |
|
View Full Product Details