Gene Transcripts
- Gene Symbol
- INTS7
- Entrez Gene ID
- 25896
- Gene Name
- integrator complex subunit 7
- Gene Aliases
- C1orf73, INT7
- Chromosome Location
- Chr. 1: 211940328 - 212035660 on Build GRCh38
- UniGene ID
- Hs.369285
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199809.1 |
NP_001186738.1
|
4, 5 |
640 |
|
NM_001199811.1 |
NP_001186740.1
|
5, 6 |
787 |
|
NM_001199812.1 |
NP_001186741.1
|
5, 6 |
787 |
|
NM_015434.3 |
NP_056249.1
|
5, 6 |
787 |
|
NR_037667.1 |
|
6, 7 |
821 |
|
XM_011509396.2 |
XP_011507698.1
|
5, 6 |
672 |
|
XM_017000962.1 |
XP_016856451.1
|
5, 6 |
672 |
|
GenBank mRNA |
AK001363.1 |
- |
5, 6 |
647 |
|
AK001598.1 |
- |
5, 6 |
669 |
|
AK022509.1 |
- |
5, 6 |
647 |
|
AK297225.1 |
- |
4, 5 |
519 |
|
BC020523.2 |
- |
5, 6 |
663 |
3917030 |
BC030716.1 |
- |
5, 6 |
642 |
3909231 |
BC033918.1 |
- |
5, 6 |
646 |
3920829 |
View Full Product Details