Gene Transcripts

Gene Symbol
HINFP
Entrez Gene ID
25988
Gene Name
histone H4 transcription factor
Gene Aliases
HiNF-P, MIZF, ZNF743
Chromosome Location
Chr. 11: 119121523 - 119135055 on Build GRCh38
UniGene ID
Hs.504091
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243259.1 NP_001230188.1 5 710
NM_015517.4 NP_056332.2 6 827
NM_198971.2 NP_945322.1 5 710
XM_011542744.2 XP_011541046.1 5 819
XM_011542745.2 XP_011541047.1 5 669
XM_011542746.1 XP_011541048.1 4 490
XM_017017499.1 XP_016872988.1 5 767
XM_017017500.1 XP_016872989.1 5 646
XM_017017501.1 XP_016872990.1 6 773
XM_017017502.1 XP_016872991.1 8 1098
XM_017017503.1 XP_016872992.1 6 863
XM_017017504.1 XP_016872993.1 6 738
XM_017017505.1 XP_016872994.1 5 661
XM_017017506.1 XP_016872995.1 5 624
XM_017017507.1 XP_016872996.1 7 863
XM_017017508.1 XP_016872997.1 6 738
XM_017017509.1 XP_016872998.1 4 490
GenBank mRNA AK056362.1 - 5 623
AK092869.1 - 1 451
AK301452.1 - 5 617
BC001073.2 - 5 681 2820431
BC001945.1 - 5 681 2820431
BC012856.1 - 5 622 3871231
BC017234.2 - 5 630 3537381

View Full Product Details