Gene Transcripts

Gene Symbol
G0S2
Entrez Gene ID
50486
Gene Name
G0/G1 switch 2
Gene Aliases
Chromosome Location
Chr. 1: 209675325 - 209676390 on Build GRCh38
UniGene ID
Hs.432132
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015714.3 NP_056529.1 2 738
GenBank mRNA AV709160.1 - 1 153
BC009694.1 - 2 642 3891372

View Full Product Details