Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297746.1 | NP_001284675.1 | 3 | 405 | |
NM_016076.4 | NP_057160.2 | 4 | 504 | ||
NR_123735.1 | 4 | 499 | |||
NR_123736.1 | 3 | 410 | |||
XM_011544203.2 | XP_011542505.1 | 5 | 589 | ||
XM_011544205.2 | XP_011542507.1 | 4 | 1155 | ||
XM_011544206.2 | XP_011542508.1 | 4 | 228 | ||
XM_011544207.2 | XP_011542509.1 | 3 | 1595 | ||
GenBank mRNA | AF151904.1 | - | 4 | 296 | |
AF229834.1 | - | 3 | 146 | ||
AK314664.1 | - | 4 | 474 | ||
BC004485.2 | - | 4 | 451 | 3831754 | |
BI459676.1 | - | 4 | 507 | ||
CB854052.1 | - | 3 | 162 | ||
CN303012.1 | - | 4 | 410 |